siRNA Details
Gene Symbol: Cml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236785.2 | XP_006236847.1 | 2 | 244 | ||
XM_006236786.2 | XP_006236848.1 | 2 | 270 | |||
XM_006236787.2 | XP_006236849.1 | 2 | 254 |
Gene Symbol: LOC100910829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236799.2 | XP_006236861.1 | 2 | 155 | ||
XM_006236800.2 | XP_006236862.1 | 154 | ||||
XM_006236801.2 | XP_006236863.1 | 140 |
Gene Symbol: LOC102549803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236818.2 | XP_006236880.1 | 1 | 1504 | ||
XM_008763111.1 | XP_008761333.1 | 2 | 1084 | |||
XM_008763112.1 | XP_008761334.1 | 2 | 1311 | |||
XM_008763113.1 | XP_008761335.1 | 2 | 687 |
Gene Symbol: LOC102556148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236802.2 | XP_006236864.1 | 687 | |||
XM_008763098.1 | XP_008761320.1 | 1 | 2721 | |||
XM_008763099.1 | XP_008761321.1 | 2 | 2301 | |||
XM_008763100.1 | XP_008761322.1 | 1311 |
