siRNA Details
Gene Symbol: LOC102556148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236802.2 | XP_006236864.1 | 408 | |||
XM_008763100.1 | XP_008761322.1 | 408 |
Gene Symbol: LOC102549803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763112.1 | XP_008761334.1 | 1 | 408 | ||
XM_008763113.1 | XP_008761335.1 | 1 | 408 |
