siRNA Details
Gene Symbol: Hmces | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025047.1 | NP_001020218.1 | 4 | 541 | ||
XM_006236851.2 | XP_006236913.1 | 5 | 925 | |||
GenBank mRNA | BC083690.1 | - | 4 | 541 | 7189409 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_173737, AK010550, BC024401, BC064070.
Gene Ontology Categories:
