siRNA Details
Gene Symbol: Dnajb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109248.1 | NP_001102718.1 | 1 | 811 | ||
XM_008763077.1 | XP_008761299.1 | 1 | 846 | |||
XM_008763078.1 | XP_008761300.1 | 1 | 846 |
