siRNA Details
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109249.1 | NP_001102719.1 | 1 | 195 | ||
XM_006236864.2 | XP_006236926.1 | 1 | 209 | |||
XM_008763079.1 | XP_008761301.1 | 1 | 210 | |||
GenBank mRNA | BC168694.1 | - | 1 | 172 | 7937199 |
