siRNA Details
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109249.1 | NP_001102719.1 | 2, 3 | 513 | ||
XM_006236864.2 | XP_006236926.1 | 2, 3 | 527 | |||
XM_008763079.1 | XP_008761301.1 | 2, 3 | 528 | |||
GenBank mRNA | BC168694.1 | - | 2, 3 | 490 | 7937199 |
