siRNA Details
Gene Symbol: Magi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030045.1 | NP_001025216.1 | 4 | 591 | ||
XM_006236927.2 | XP_006236989.1 | 4 | 608 | |||
XM_006236928.2 | XP_006236990.1 | 4 | 608 | |||
XM_006236929.2 | XP_006236991.1 | 4 | 608 | |||
XM_006236930.2 | XP_006236992.1 | 4 | 608 | |||
XM_006236931.2 | XP_006236993.1 | 4 | 608 | |||
XM_008763131.1 | XP_008761353.1 | 4 | 608 | |||
XM_008763132.1 | XP_008761354.1 | 4 | 608 | |||
XM_008763133.1 | XP_008761355.1 | 4 | 608 | |||
GenBank mRNA | AY598952.1 | - | 4 | 591 |
