siRNA Details
Gene Symbol: Lpar5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749835.3 | XP_003749883.1 | 2 | 1352 | ||
XM_006237389.2 | XP_006237451.1 | 3 | 1472 | |||
XM_006237391.2 | XP_006237453.1 | 3 | 1287 | |||
XM_006237392.2 | XP_006237454.1 | 3 | 1443 | |||
XM_008763296.1 | XP_008761518.1 | 4 | 1378 | |||
XM_008763297.1 | XP_008761519.1 | 2 | 1323 | |||
XM_008763298.1 | XP_008761520.1 | 2 | 1272 | |||
XM_575667.6 | XP_575667.2 | 2 | 1167 |
