siRNA Details
Gene Symbol: Parp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237510.2 | XP_006237572.1 | 4 | 1624 | ||
XM_008763302.1 | XP_008761524.1 | 8 | 790 | |||
XM_008763303.1 | XP_008761525.1 | 7 | 692 | |||
XM_008763304.1 | XP_008761526.1 | 7 | 807 | |||
XM_008763305.1 | XP_008761527.1 | 6 | 619 |
