siRNA Details
Gene Symbol: Rergl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191979.1 | NP_001178908.1 | 5 | 556 | ||
XM_008763418.1 | XP_008761640.1 | 4 | 552 |

More Information
Other Species Targeted

Gene Ontology Categories:
