siRNA Details
Gene Symbol: Lmntd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763419.1 | XP_008761641.1 | 3, 4 | 1650 | ||
XM_008763420.1 | XP_008761642.1 | 2, 3 | 215 | |||
XM_008763421.1 | XP_008761643.1 | 1, 2 | 259 |
Gene Symbol: LOC100912509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237675.2 | XP_006237737.1 | 1, 2 | 353 |
