siRNA Details
Gene Symbol: Tuba8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024339.1 | NP_001019510.1 | 4 | 1005 | ||
XM_008763254.1 | XP_008761476.1 | 4 | 1142 | |||
GenBank mRNA | BC079185.1 | - | 4 | 1005 | 7131106 | |
FQ217798.1 | - | 4 | 1061 |
