siRNA Details
Gene Symbol: Rmdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031663.1 | NP_001026833.1 | 6, 7 | 642 | ||
XM_006237933.2 | XP_006237995.1 | 5, 6 | 685 | |||
GenBank mRNA | BC079473.1 | - | 6, 7 | 648 | ||
BC098713.1 | - | 6, 7 | 642 | 7322407 |
