siRNA Details
Gene Symbol: RGD1563056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238003.2 | XP_006238065.1 | 5 | 471 | ||
XM_008763643.1 | XP_008761865.1 | 4 | 1530 | |||
XM_575796.4 | XP_575796.2 | 4 | 1091 |
