siRNA Details
Gene Symbol: Sit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024344.1 | NP_001019515.1 | 5 | 552 | ||
GenBank mRNA | BC088199.1 | - | 5 | 552 | 7377221 | |
FQ225852.1 | - | 4 | 467 | |||
FQ233921.1 | - | 4 | 467 |
