siRNA Details
Gene Symbol: Caap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034154.1 | NP_001029326.1 | 6 | 624 | ||
XM_006238400.2 | XP_006238462.1 | 6 | 838 | |||
XM_008763894.1 | XP_008762116.1 | 6 | 622 | |||
GenBank mRNA | BC105829.1 | - | 6 | 624 | 7382013 |
