siRNA Details
Gene Symbol: RGD1562532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238487.2 | XP_006238549.1 | 3 | 209 | ||
XM_008763920.1 | XP_008762142.1 | 2 | 656 | |||
XM_575875.6 | XP_575875.4 | 3 | 747 | |||
GenBank mRNA | BC079057.1 | - | 3 | 470 | ||
BC105617.1 | - | 1 | 71 |
