siRNA Details
Gene Symbol: RGD1559493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109262.2 | NP_001102732.1 | 10 | 1225 | ||
GenBank mRNA | BC160900.1 | - | 10 | 1225 | 7454065 |

More Information
Other Species Targeted

