siRNA Details
Gene Symbol: Mknk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044267.1 | NP_001037732.1 | 3 | 231 | ||
XM_006238694.2 | XP_006238756.1 | 3 | 1777 | |||
XM_006238695.2 | XP_006238757.1 | 3 | 1779 | |||
XM_006238696.2 | XP_006238758.1 | 6 | 592 | |||
XM_006238697.2 | XP_006238759.1 | 5 | 544 | |||
XM_006238698.2 | XP_006238760.1 | 5 | 485 | |||
XM_006238699.2 | XP_006238761.1 | 3 | 1711 | |||
GenBank mRNA | BC098754.1 | - | 3 | 231 | 7370127 |
