siRNA Details
Gene Symbol: Mknk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044267.1 | NP_001037732.1 | 6 | 484 | ||
XM_006238694.2 | XP_006238756.1 | 6 | 2030 | |||
XM_006238695.2 | XP_006238757.1 | 6 | 2032 | |||
XM_006238696.2 | XP_006238758.1 | 9 | 845 | |||
XM_006238697.2 | XP_006238759.1 | 8 | 797 | |||
XM_006238698.2 | XP_006238760.1 | 8 | 738 | |||
XM_006238699.2 | XP_006238761.1 | 6 | 1964 | |||
GenBank mRNA | BC098754.1 | - | 6 | 484 | 7370127 |
