siRNA Details
Gene Symbol: Cdca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025050.1 | NP_001020221.1 | 2 | 332 | ||
XM_008764064.1 | XP_008762286.1 | 3 | 332 | |||
GenBank mRNA | BC079274.1 | - | 2 | 332 | 7134027 | |
BC079293.1 | - | 3 | 311 | 7134482 |
