siRNA Details
Gene Symbol: RGD1560436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109264.1 | NP_001102734.1 | 2 | 611 | ||
XM_006238898.2 | XP_006238960.1 | 2 | 658 | |||
XM_006238900.2 | XP_006238962.1 | 3 | 559 | |||
XM_008764065.1 | XP_008762287.1 | 2 | 661 |
