siRNA Details
Gene Symbol: S100pbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134628.1 | NP_001128100.1 | 7 | 1387 | ||
XM_006238956.2 | XP_006239018.1 | 7 | 1354 | |||
XM_006238957.2 | XP_006239019.1 | 7 | 1658 | |||
XM_006238958.2 | XP_006239020.1 | 7 | 1370 | |||
XM_006238959.2 | XP_006239021.1 | 6 | 1240 | |||
XM_006238960.1 | XP_006239022.1 | 6 | 1299 | |||
XM_006238961.2 | XP_006239023.1 | 8 | 1228 | |||
XM_006238963.2 | XP_006239025.1 | 8 | 1124 |
