siRNA Details
Gene Symbol: S100pbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134628.1 | NP_001128100.1 | 3 | 758 | ||
XM_006238956.2 | XP_006239018.1 | 3 | 725 | |||
XM_006238957.2 | XP_006239019.1 | 3 | 1029 | |||
XM_006238958.2 | XP_006239020.1 | 3 | 741 | |||
XM_006238959.2 | XP_006239021.1 | 2 | 611 | |||
XM_006238960.1 | XP_006239022.1 | 2 | 670 | |||
XM_006238961.2 | XP_006239023.1 | 3 | 725 | |||
XM_006238962.2 | XP_006239024.1 | 3 | 724 | |||
XM_006238963.2 | XP_006239025.1 | 4 | 495 | |||
XM_008764162.1 | XP_008762384.1 | 3 | 725 | |||
XM_008764163.1 | XP_008762385.1 | 3 | 724 | |||
XM_008764164.1 | XP_008762386.1 | 3 | 724 |
