siRNA Details
Gene Symbol: Gmeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109268.2 | NP_001102738.1 | 3, 4 | 412 | ||
XM_008764165.1 | XP_008762387.1 | 3, 4 | 745 | |||
XM_008764166.1 | XP_008762388.1 | 4, 5 | 1108 | |||
XM_008764167.1 | XP_008762389.1 | 4, 5 | 1105 | |||
XM_008764168.1 | XP_008762390.1 | 5, 6 | 996 | |||
XM_008764169.1 | XP_008762391.1 | 4, 5 | 939 | |||
GenBank mRNA | BC158640.1 | - | 3, 4 | 412 | 8362091 |
