siRNA Details
Gene Symbol: Tmco4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034949.1 | NP_001030121.1 | 12 | 1462 | ||
XM_006239262.1 | XP_006239324.1 | 12 | 2887 | |||
GenBank mRNA | BC099755.1 | - | 12 | 1462 | 5623720 |
