siRNA Details
Gene Symbol: Szrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114599.2 | NP_001108071.1 | 4 | 404 | ||
XM_006239264.2 | XP_006239326.1 | 4 | 432 | |||
GenBank mRNA | BC083784.1 | - | 5 | 486 | ||
FQ213371.1 | - | 5 | 505 | |||
FQ222061.1 | - | 3 | 340 |
