siRNA Details
Gene Symbol: Szrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114599.2 | NP_001108071.1 | 2, 3 | 129 | ||
GenBank mRNA | BP502965.1 | - | 2, 3 | 116 | ||
FQ213371.1 | - | 2, 3 | 127 | |||
FQ222061.1 | - | 1, 2 | 65 |
