siRNA Details
Gene Symbol: Dcdc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195807.1 | NP_001182736.1 | 7 | 837 | ||
XM_006239955.2 | XP_006240017.1 | 7 | 1208 | |||
XM_006239956.2 | XP_006240018.1 | 7 | 1207 | |||
XM_006239957.2 | XP_006240019.1 | 7 | 1207 | |||
XM_006239958.2 | XP_006240020.1 | 7 | 1207 |
