siRNA Details
Gene Symbol: Ccdc71l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750141.3 | XP_003750189.2 | 1 | 2802 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001162903, AK171109.
