siRNA Details
Gene Symbol: Sptssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047743.1 | NP_001041208.1 | 1, 2 | 113 | ||
GenBank mRNA | BC098829.1 | - | 1, 2 | 113 |

More Information
Other Species Targeted

Gene Ontology Categories:
