siRNA Details
Gene Symbol: Ccdc175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164400.1 | NP_001157872.1 | 11, 12 | 1364 | ||
XM_008764718.1 | XP_008762940.1 | 11, 12 | 1407 | |||
GenBank mRNA | BC087160.1 | - | 11, 12 | 1364 | 7134746 |
