siRNA Details
Gene Symbol: Plekhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127566.1 | NP_001121038.1 | 9 | 1048 | ||
GenBank mRNA | BC161896.1 | - | 9 | 1048 | 7319876 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plekhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127566.1 | NP_001121038.1 | 9 | 1048 | ||
GenBank mRNA | BC161896.1 | - | 9 | 1048 | 7319876 |