siRNA Details
Gene Symbol: Plekhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127566.1 | NP_001121038.1 | 11 | 1329 | ||
GenBank mRNA | BC161896.1 | - | 11 | 1329 | 7319876 |

More Information
Other Species Targeted

