siRNA Details
Gene Symbol: Efcab11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024350.1 | NP_001019521.1 | 1, 2 | 135 | ||
XM_006240446.2 | XP_006240508.2 | 1, 2 | 237 | |||
GenBank mRNA | BC079408.1 | - | 1, 2 | 135 | 7113005 |
