siRNA Details
Gene Symbol: Tmem198b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109281.1 | NP_001102751.1 | 4, 5 | 1404 | ||
XM_006240794.2 | XP_006240856.1 | 4, 5 | 1265 | |||
XM_006240795.1 | XP_006240857.1 | 3, 4 | 1319 | |||
XM_008765038.1 | XP_008763260.1 | 3, 4 | 1709 |
