siRNA Details
Gene Symbol: Ccdc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241271.2 | XP_006241333.1 | 7 | 666 | ||
XM_008765310.1 | XP_008763532.1 | 6 | 396 | |||
XM_008765311.1 | XP_008763533.1 | 7 | 678 |
