siRNA Details
Gene Symbol: Fgd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137645.1 | NP_001131117.1 | 8, 9 | 3314 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fgd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137645.1 | NP_001131117.1 | 8, 9 | 3314 |