siRNA Details
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241616.1 | XP_006241678.1 | 4 | 549 | ||
XM_006241617.1 | XP_006241679.1 | 4 | 549 | |||
XM_006241618.2 | XP_006241680.1 | 3 | 709 | |||
XM_008765476.1 | XP_008763698.1 | 3 | 490 | |||
XM_008765477.1 | XP_008763699.1 | 3 | 493 | |||
GenBank mRNA | FQ211706.1 | - | 4 | 549 |
