siRNA Details
Gene Symbol: Mtbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130717.1 | NP_001124189.1 | 15 | 1966 | ||
XM_006241655.1 | XP_006241717.1 | 15 | 1646 | |||
XM_006241656.2 | XP_006241718.1 | 15 | 1646 | |||
XM_008765516.1 | XP_008763738.1 | 15 | 1646 | |||
XM_008765517.1 | XP_008763739.1 | 15 | 1646 |
