siRNA Details
Gene Symbol: RGD1562063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241913.2 | XP_006241975.1 | 11 | 2590 | ||
XM_006241914.2 | XP_006241976.1 | 9 | 2335 | |||
XM_008765632.1 | XP_008763854.1 | 11 | 2590 |
