siRNA Details
Gene Symbol: Lrrc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024354.1 | NP_001019525.1 | 3 | 707 | ||
XM_006241860.2 | XP_006241922.1 | 3 | 962 | |||
XM_006241861.1 | XP_006241923.1 | 3 | 595 | |||
XM_006241862.2 | XP_006241924.1 | 4 | 1480 | |||
XM_006241865.1 | XP_006241927.1 | 4 | 690 | |||
GenBank mRNA | BC092583.1 | - | 3 | 707 | 7376401 | |
FQ213127.1 | - | 1 | 97 |
