siRNA Details
Gene Symbol: Ncf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127304.1 | NP_001120776.1 | 10 | 1077 | ||
XM_006241939.2 | XP_006242001.1 | 11 | 1741 | |||
GenBank mRNA | BC167076.1 | - | 10 | 1049 | 7129758 |
