siRNA Details
Gene Symbol: Arhgap42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242515.2 | XP_006242577.1 | 2 | 965 | ||
XM_008765909.1 | XP_008764131.1 | 2 | 1418 | |||
XM_576354.6 | XP_576354.3 | 2 | 508 |
