siRNA Details
Gene Symbol: LOC500948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047964.1 | NP_001041429.1 | 443 | |||
GenBank mRNA | AY387059.1 | - | 443 |
Gene Symbol: LOC103693015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766000.1 | XP_008764222.1 | 3 | 1174 | ||
XM_008766001.1 | XP_008764223.1 | 2 | 1064 |
