siRNA Details
Gene Symbol: Ubl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048243.2 | NP_001041708.2 | 3 | 188 | ||
XM_006242661.2 | XP_006242723.1 | 3 | 369 | |||
GenBank mRNA | BC091358.1 | - | 2 | 382 | ||
BC157818.1 | - | 2 | 91 | 7936655 | ||
CB713235.1 | - | 3 | 188 |
