siRNA Details
Gene Symbol: RGD1560775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766460.1 | XP_008764682.1 | 20, 21 | 1625 | ||
XM_008766461.1 | XP_008764683.1 | 20, 21 | 1364 | |||
XM_008766462.1 | XP_008764684.1 | 20, 21 | 1625 | |||
XM_008766463.1 | XP_008764685.1 | 20, 21 | 1625 | |||
XM_008766464.1 | XP_008764686.1 | 19, 20 | 1512 | |||
XM_008766465.1 | XP_008764687.1 | 19, 20 | 1433 | |||
GenBank mRNA | BC128758.1 | - | 19, 20 | 1207 | 7111588 |
