siRNA Details
Gene Symbol: Golga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191080.1 | NP_001178009.1 | 14 | 4450 | ||
XM_006244126.2 | XP_006244188.1 | 16 | 4970 | |||
XM_006244127.2 | XP_006244189.1 | 16 | 4940 | |||
XM_006244128.2 | XP_006244190.1 | 16 | 4701 | |||
XM_006244129.2 | XP_006244191.1 | 16 | 4970 | |||
XM_006244130.2 | XP_006244192.1 | 15 | 4886 | |||
XM_006244131.2 | XP_006244193.1 | 15 | 4871 | |||
XM_006244132.2 | XP_006244194.1 | 16 | 4970 | |||
XM_006244133.2 | XP_006244195.1 | 15 | 4871 |
