siRNA Details
Gene Symbol: Rfx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767059.1 | XP_008765281.1 | 14 | 2695 | ||
GenBank mRNA | FQ230624.1 | - | 9 | 1256 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rfx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767059.1 | XP_008765281.1 | 14 | 2695 | ||
GenBank mRNA | FQ230624.1 | - | 9 | 1256 |