siRNA Details
Gene Symbol: Zdbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245044.2 | XP_006245106.1 | 4 | 3637 | ||
XM_006245045.2 | XP_006245107.1 | 4 | 3620 | |||
XM_006245046.2 | XP_006245108.1 | 4 | 3667 | |||
XM_008767157.1 | XP_008765379.1 | 5 | 3939 | |||
XM_008767158.1 | XP_008765380.1 | 5 | 3960 | |||
XM_008767159.1 | XP_008765381.1 | 6 | 4022 | |||
XM_576579.7 | XP_576579.4 | 5 | 4646 |
